Kaliumbromid, Premium FTIR-Qualität
Lieferant: Thermo Fisher Scientific
|
Achtung
|
47111.KD
47111.KM
47111.KN
47111.KQ
47111.KDEA
208
EUR
47111.KD
47111.KM
47111.KN
47111.KQ
Kaliumbromid, Premium FTIR-Qualität
Kaliumbromid
CAS No.: 7758-02-3
Molecular Formula: BrK
Formula Weight: 119.01
Physical Form: Powder in 0.5g Packets
Density: 2.75
Refractive Index: 1.559
Odour: Odourless
Melting Point: 730°
Boiling Point: 1435°
MDL No.: MFCD00011358
Formel:
KBr Siedepunkt: 1435 °C Schmelzpunkt: 730 °C Dichte: 2,75 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011358 CAS-Nummer: 7758-02-3 EINECS: 231-830-3 Merck Index: 14,7618 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...