Maraviroc 97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
UK427857, MVC, 4,4-Difluoro-N-((S)-3-((1R,3R,5S)-3-(3-isopropyl-5-methyl-4H-1,2,4-triazol-4-yl)-8-azabicyclo[3.2.1]octan-8-yl)-1-phenylpropyl)cyclohexane-1-carboxamide, 4,4-difluoro-N-[(1S)-3-[(3-exo)-3-[3-methyl-5-(1-methylethyl)-4H-1,2,4-triazol-4-yl]-8-azabicyclo[3.2.1]oct-8-yl]-1-phenylpropyl]-cyclohexanecarboxamide
471172500
471172500.EA
404
EUR
471172500.
Maraviroc 97%
Maraviroc
Formel:
C₂₉H₄₁F₂N₅O Molecular Weight: 513,67 g/mol Lagertemperatur: Gefrierschrank |
MDL:
MFCD13188530 CAS-Nummer: 376348-65-1 |
Spezifikation Testergebnisse
Appearance | white to almost white powder |
Infrared spectrum | Conforms |
HPLC | ≥96 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...