Olivenöl

Lieferant: Merck

47118
47118.EA 87 EUR
47118.
Olivenöl
Olive oil
MDL: MFCD00131764
CAS-Nummer: 8001-25-0
EINECS: 232-277-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR