Bariumiodid, ultra trocken ≥99,999% (Metall-Basis), Perlen -10 Mesh

Lieferant: Thermo Scientific
Achtung


Unter Argon in Ampullen verpackt

47118.14
47118.14EA 582 EUR
47118.14
Bariumiodid, ultra trocken ≥99,999% (Metall-Basis), Perlen -10 Mesh
Bariumiodid

Barium iodide is used as a reagent for facile synthesis of poly(phenylcarbyne). It reacts with alkyl potassium compounds to form organobarium compounds.


Purity: 99.999% CAS No.: 13718-50-8 Molecular Formula: BaI2 Formula Weight: 391.15 Physical Form: -10 Mesh Beads, Ampouled under argon Density: 5.15 Sensitivity: Light Sensitive; Hygroscopic Appearance: White Odour: Odourless Melting Point: 740° UN No.: UN1564 MDL No.: MFCD00003452

Formel: BaI₂
Molecular Weight: 391,14 g/mol
Schmelzpunkt: 740 °C
Dichte: 5,15
Lagertemperatur: Raumtemperatur
MDL: MFCD00003452
CAS-Nummer: 13718-50-8
EINECS: 237-276-9
UN: 1564
ADR: 6.1,III
Merck Index: 14,980

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR