Resiquimod 98%

Lieferant: Thermo Scientific

Synonyme: 1-[4-Amino-2-(ethoxymethyl)-1H-imidazo[4,5-c]quinolin-1-yl]-2-methylpropan-2-ol, 4-Amino-2-(ethoxymethyl)-a,a-dimethyl-1H-imidazo[4,5-c]quinoline-1-ethanol, 1-(4-Amino-2-(ethoxymethyl)-1H-imidazo[4,5-c]quinolin-1-yl)-2-methylpropan-2-ol, 4-Amino-2-(ethoxymethyl)-alpha,alpha-dimethyl-1H-imidazo[4,5-c]quinoline-1-ethanol, R-848

471212500
471212500.EA 187 EUR
471212500.
Resiquimod 98%
Resiquimod
Formel: C₁₇H₂₂N₄O₂
Molecular Weight: 314,389 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00937759
CAS-Nummer: 144875-48-9

Bestellen

Spezifikation Testergebnisse

Appearance white to yellow to beige crystalline powder
Infrared spectrum Conforms
HPLC ≥97.5 %
Melting point 190°C to 195°C
Water ≤2 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR