Quercitrin 98%

Lieferant: Thermo Scientific

Synonyme: Quercetin 3-rhamnoside, 2-(3,4-Dihydroxyphenyl)-5,7-dihydroxy-3-(((2S,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyltetrahydro-2H-pyran-2-yl)oxy)-4H-chromen-4-one

471311000
471311000.EA 177 EUR
471311000.
Quercitrin 98%
Quercitrin
Formel: C₂₁H₂₀O₁₁
Molecular Weight: 448,38 g/mol
Siedepunkt: 814°C
Lagertemperatur: Kühlschrank
MDL: MFCD00016932
CAS-Nummer: 522-12-3
EINECS: 208-322-5

Bestellen

Spezifikation Testergebnisse

Appearance white to yellow crystalline powder
Infrared spectrum Conforms
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR