Paeoniflorin 95%

Lieferant: Thermo Scientific
Achtung

Synonyme: ((2S,2aR,2a1S,3aR,4R,5aR)-4-Hydroxy-2-methyl-2a-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)hexahydro-2H-1,5-dioxa-2,4-methanocyclobuta[cd]pentalen-2a1-yl)methyl benzoate

471331000
471331000.EA 82.2 EUR
471331000.
Paeoniflorin 95%
Paeoniflorin
Formel: C₂₃H₂₈O₁₁
Molecular Weight: 480,47 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00869331
CAS-Nummer: 23180-57-6
EINECS: 245-476-2

Bestellen

Spezifikation Testergebnisse

Appearance white powder
Infrared spectrum Conforms
HPLC ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR