Wogonin 98%
Lieferant: Thermo Scientific
Synonyme:
5,7-Dihydroxy-8-methoxy-2-phenyl-4H-chromen-4-one, 5,7-Dihydroxy-8-methoxyflavone
471341000
471341000.EA
145
EUR
471341000.
Wogonin 98%
Wogonin
Formel:
C₁₆H₁₂O₅ Molecular Weight: 284,3 g/mol Siedepunkt: 518,8°C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00017736 CAS-Nummer: 632-85-9 |
Spezifikation Testergebnisse
Appearance | white to beige crystalline powder |
Infrared spectrum | Conforms |
HPLC | ≥97.5 % |
Melting point | 205°C to 209°C |
Loss on drying | ≤3 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...