Vitexin 97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 8-Glucopyranosylapigenin, 5,7-Dihydroxy-2-(4-hydroxyphenyl)-8-((2S,3R,4R,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)-4H-chromen-4-one

471351000
471351000.EA 274 EUR
471351000.
Vitexin 97%
Vitexin
Formel: C₂₁H₂₀O₁₀
Molecular Weight: 432.4 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00017456
CAS-Nummer: 3681-93-4
EINECS: 222-963-8

Bestellen

Spezifikation Testergebnisse

Appearance yellow powder
Infrared spectrum Conforms
HPLC ≥96 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR