Spiro-OMeTAD 98%
Lieferant: Thermo Scientific
Synonyme:
2,2',7,7'-Tetrakis[N,N-di(4-methoxyphenyl)amino]-9,9'-spirobifluorene, N2,N2,N2',N2',N7,N7,N7',N7'-Octakis(4-methoxyphenyl)-9,9'-spirobi[fluorene]-2,2',7,7'-tetraamine
471370010
471370010.EA
282
EUR
471370010.
Spiro-OMeTAD 98%
Spiro-OMeTAD
Formel:
C₈₁H₆₈N₄O₈ Molecular Weight: 1225,42 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD12022511 CAS-Nummer: 207739-72-8 |
Spezifikation Testergebnisse
Appearance | white to light green to yellow crystalline powder |
Infrared spectrum | Conforms |
HPLC | ≥97.5 % |
Melting point | 245°C to 250°C |
UV Lambda max | 290 to 310 nm (c=10, CH2Cl2) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...