Tin bis(trifluoromethylsulphonyl)imide
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
47142.MD
47142.03
47142.06
47142.MDEA
47.3
EUR
47142.MD
47142.03
47142.06
Tin bis(trifluoromethylsulphonyl)imide
Tin bis(trifluoromethylsulphonyl)imide
Molecular Formula: C4F12N2SnO8S4
Formula Weight: 678.98
Physical Form: Solid
Sensitivity: Hygroscopic
Appearance: Off-white
UN No.: UN3261
MDL No.: MFCD28100488
Formel:
C₄F₁₂N₂SnO₈S₄ Lagertemperatur: Raumtemperatur |
MDL:
MFCD28100488 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...