Sophoricoside 98%

Lieferant: Thermo Fisher Scientific

Synonyme: Genistein 4'-Glucoside, 5,7-Dihydroxy-3-(4-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)phenyl)-4H-chromen-4-one, 4'-(beta-D-Glucopyranosyloxy)-5,7-dihydroxyisoflavone

471420010 471420050
471420010.EA 86.2 EUR
471420010. 471420050.
Sophoricoside 98%
Sophoricoside
Formel: C₂₁H₂₀O₁₀
Lagertemperatur: Raumtemperatur
MDL: MFCD01075138
CAS-Nummer: 152-95-4

Bestellen

Spezifikation Testergebnisse

Appearance white to light yellow crystalline powder
Infrared spectrum Conforms
HPLC ≥97.5 %
Loss on drying ≤3 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR