Cobalt, oil based standard solution, Co 1000 µg/g, Specpure®
Lieferant: Thermo Fisher Scientific
47143.18
47143.18EA
108
EUR
47143.18
Cobalt, oil based standard solution, Co 1000 µg/g, Specpure®
Standards
Kationischer Standards
Cobalt-Standards
Physical Form: Liquid
MDL No.: MFCD00010935
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...