Physcion 98%

Lieferant: Thermo Scientific
Achtung

Synonyme: Emodin-3-methyl ether, 1,8-Dihydroxy-3-methoxy-6-methyl-anthrachinon, 1,8-Dihydroxy-3-methoxy-6-methylanthracene-9,10-dione, Parietin

471431000
471431000.EA 191 EUR
471431000.
Physcion 98%
Physcion
Formel: C₁₆H₁₂O₅
Molecular Weight: 284,28 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00017374
CAS-Nummer: 521-61-9
EINECS: 208-315-7

Bestellen

Spezifikation Testergebnisse

Appearance yellow to orange crystalline powder
Infrared spectrum Conforms
HPLC ≥97.5 %
Melting point 200°C to 205°C
Water ≤3 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR