Iridium, schwarz ≥99,95% (Metall-Basis)

Lieferant: Thermo Fisher Scientific
Achtung

47150.ME 47150.04 47150.09
47150.MEEA 408 EUR
47150.ME 47150.04 47150.09
Iridium, schwarz ≥99,95% (Metall-Basis)
Iridium

Purity: 99.95% CAS No.: 7439-88-5 Physical Form: S.A. 20-30m2/g Appearance: Black UN No.: UN3089 MDL No.: MFCD00011062

Formel: Ir
Lagertemperatur: Raumtemperatur
MDL: MFCD00011062
CAS-Nummer: 7439-88-5
EINECS: 231-095-9
UN: 3089
ADR: 4.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR