Phosphorus, plasma standard solution, P 1,0000 µg/ml, Specpure®
Lieferant: Thermo Fisher Scientific
47152.AE
47152.AP
47152.AEEA
139
EUR
47152.AE
47152.AP
Phosphorus, plasma standard solution, P 1,0000 µg/ml, Specpure®
Standards
Kationischer Standards
Phosphor-Standards
Molecular Formula: H₃PO₄, 5% HNO₃
Physical Form: Liquid
UN No.: UN3264
MDL No.: MFCD00133771
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...