Mangan(II)iodid, ultra trocken ≥99,99% (Metall-Basis)
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Unter Argon in Ampullen verpackt
Purity: 99.99%
CAS No.: 7790-33-2
Molecular Formula: MnI2
Formula Weight: 308.75
Physical Form: -10 Mesh Beads, Ampouled under argon
Density: 5.0
Sensitivity: Light, Air and Moisture Sensitive
Appearance: White to pink to red
Odour: Odourless
Melting Point: 638°
MDL No.: MFCD00016225
Formel:
I₂Mn Schmelzpunkt: 638 °C Dichte: 5,0 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00016225 CAS-Nummer: 7790-33-2 EINECS: 232-201-6 Merck Index: 14,5732 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...