Aluminiumoxid ≥99%

Lieferant: Thermo Fisher Scientific

Synonyme: Aluminium(III) oxide, Alumina C, Aluminum oxide

47171.A3 47171.36
47171.A3EA 330 EUR
47171.A3 47171.36
Aluminiumoxid ≥99%
Aluminium oxide

Purity: 99% CAS No.: 1344-28-1 Molecular Formula: Al2O3 Formula Weight: 101.96 Physical Form: 32-63 Micron APS Powder, S.A. 200m2/g Density: 3.965 Refractive Index: 1.768 Appearance: White Odour: Odourless Melting Point: 2045° Boiling Point: 2980° MDL No.: MFCD00003424

Formel: Al₂O₃
Molecular Weight: 101.96 g/mol
Siedepunkt: 2980 °C
Schmelzpunkt: 2045 °C
Dichte: 3.965
Lagertemperatur: Raumtemperatur
MDL: MFCD00003424
CAS-Nummer: 1344-28-1
EINECS: 215-691-6
Merck Index: 14,356

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR