Quercetin-3-o-glucopyranoside 97%

Lieferant: Thermo Scientific

Synonyme: Isoquercitrin, Quercetin-3-O-glucoside, 2-(3,4-Dihydroxyphenyl)-5,7-dihydroxy-3-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-4H-chromen-4-one, Isoquercetin

471801000
471801000.EA 149 EUR
471801000.
Quercetin-3-o-glucopyranoside 97%
Quercetin-3-o-glucopyranoside
Formel: C₂₁H₂₀O₁₂
Molecular Weight: 464,376 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00017746
CAS-Nummer: 482-35-9

Bestellen

Spezifikation Testergebnisse

Appearance light yellow to amber powder
Infrared spectrum Conforms
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR