1,7-Dimethylxanthin 98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 2,6-Dihydroxy-1,7-dimethylpurin, Paraxanthin, 3,7-Dihydro-1,7-dimethyl-1H-purin-2,6-dion, 1,7-Dimethyl-3,7-dihydro-2H-purin-2,6-dion, 1,7-Dimethyl-1H,3H-purin-2,6-dion, 1,7-Dimethyl-2,6(1H,3H)-purindion, 1,7-Dimethyl-3,7-dihydro-1H-purin-2,6-dion

471832500
471832500.EA 238 EUR
471832500.
1,7-Dimethylxanthin 98%
1,7-Dimethylxanthin
Formel: C₇H₈N₄O₂
Molecular Weight: 180.17 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00005727
CAS-Nummer: 611-59-6
EINECS: 210-271-9

Bestellen

Spezifikation Testergebnisse

Appearance white to almost white powder
Infrared spectrum Conforms
HPLC ≥97.5 %
Melting point 294°C to 299°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR