L-Chicoric acid 98%

Lieferant: Thermo Scientific

Synonyme: Chicoric acid, (2R,3R)-2,3-Bis[[(E)-3-(3,4-dihydroxyphenyl)acryloyl]oxy]succinic acid

471921000
471921000.EA 341 EUR
471921000.
L-Chicoric acid 98%
L-Chicoric acid
Formel: C₂₂H₁₈O₁₂
Molecular Weight: 474,37 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD22683653
CAS-Nummer: 70831-56-0

Bestellen

Spezifikation Testergebnisse

Appearance white to light yellow to light beige powder or crystals
Infrared spectrum Conforms
HPLC ≥97.5 %
Titration with NaOH 97.5 to 102.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR