Tilorone dihydrochloride 95%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
2,7-Bis[2-(diethylamino)ethoxy]-9-fluorenone Dihydrochloride, 2,7-Bis(2-(Diethylamino)ethoxy)-9H-fluoren-9-one dihydrochloride
471940010
471940010.EA
190
EUR
471940010.
Tilorone dihydrochloride 95%
Tilorone dihydrochloride
Formel:
C₂₅H₃₄N₂O₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00134071 CAS-Nummer: 27591-69-1 Merck Index: 13,09517 |
Spezifikation Testergebnisse
Appearance | light yellow to orange powder, crystalline powder and/or chunks |
Infrared spectrum | Conforms |
Titration Argentometric | 94 to 106 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...