Pharmaceuticals refrigerator, MED-85
Lieferant: KIRSCH PHILIPP
10904
472-0310EA 0 EUR
472-0310
Pharmaceuticals refrigerator, MED-85
Kühlschränke
Refrigerator with outer housing made of galvanised sheet steel (rust-protected), with white, scratch-resistant powder coating. Interior made of impact-resistant, white plastic with moulded-in pads.
  • Automatic defrost with condensation water evaporation
  • Stepless temperature selection
  • Drawer division with fixed longitudinal and transverse dividers
  • Dewatering vessel for manual emptying
  • Chiller fan when static ventilation is insufficient during installation or when operating at 60 Hz
  • GSM module
  • Temperature recorder for insertion with two wax paper strips
This refrigerator features temperature sensor PT 100 or PT 1000, four-wire, class 1/3 B, including reference body (measuring bottle or cold block), installed.