Nonaethylenglykol 95%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 3,6,9,12,15,18,21,24-Octaoxahexacosane-1,26-diol, PEG-9

472010010
472010010.EA 103 EUR
472010010.
Nonaethylenglykol 95%
Nonaethylenglykol
Formel: C₁₈H₃₈O₁₀
Molecular Weight: 414.49 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00698692
CAS-Nummer: 3386-18-3
EINECS: 222-206-1

Bestellen

Spezifikation Testergebnisse

Appearance white to almost white low melting solid or liquid
Infrared spectrum Conforms
HPLC ≥94 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR