Undecaethylene glycol 95%

Lieferant: Thermo Scientific

Synonyme: 3,6,9,12,15,18,21,24,27,30-Decaoxadotriacontane-1,32-diol, PEG-11, HO-PEG11-OH

472020010
472020010.EA 207 EUR
472020010.
Undecaethylene glycol 95%
Undecaethylene glycol
Formel: C₂₂H₄₆O₁₂
Molecular Weight: 502,59 g/mol
Flammpunkt: 299°C
Lagertemperatur: Raumtemperatur
MDL: MFCD26793839
CAS-Nummer: 6809-70-7
EINECS: 229-884-8

Bestellen

Spezifikation Testergebnisse

Appearance white to light yellow low melting solid
Infrared spectrum Conforms
HPLC ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR