Ethyl 4′-hydroxy-4-biphenylcarboxylate 98, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY
Achtung

Synonyme: Ethyl-4'-hydroxy[1,1'-biphenyl]-4-carboxylate, Ethyl 4'-hydroxy-[1,1'-biphenyl]-4-carboxylate

MQ200

472069-1G 472069-5G
472069-1GEA 93.8 EUR
472069-1G 472069-5G
Ethyl 4′-hydroxy-4-biphenylcarboxylate 98, Sigma-Aldrich®
Ethyl 4′-hydroxy-4-biphenylcarboxylate

Ethyl 4'-hydroxy-4-biphenylcarboxylate,98%

Formel: C₁₅H₁₄O₃
Molecular Weight: 242.28 g/mol
Schmelzpunkt: 142…144 °C (Zers., Lit.)
Lagertemperatur: Room Temperature
MDL: MFCD07809563
CAS-Nummer: 50670-76-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ee25e187ea65ba2e10229d35e0cb961d9acda764&vid=e45fd63177b9003b592fe02856530517e12a6173&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Appearance (Colour) White to Yellow
Appearance (Form) Conforms to Requirements
Powder or Crystals
Infrared spectrum Conforms to Structure
Carbon 72.5 - 76.2 %
Purity (HPLC) ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR