2-(4-Hydroxyphenyl)-5-pyrimidinol 90%, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY
Achtung


M-Clarity™ Qualitätsstufe = MQ100

472077-1G
472077-1GEA 127 EUR
472077-1G
2-(4-Hydroxyphenyl)-5-pyrimidinol 90%, Sigma-Aldrich®
2-(4-Hydroxyphenyl)-5-pyrimidinol
Formel: C₁₀H₈N₂O₂
Molecular Weight: 188,18 g/mol
Schmelzpunkt: 235,..240 °C (Lit.)
Lagertemperatur: Room Temperature
MDL: MFCD01321266
CAS-Nummer: 142172-97-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e48ac78865fbad3243200691c10d6ddb37664798&vid=e48ac78865fbad3243200691c10d6ddb37664798&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Appearance (Colour), White to Yellow to Beige Conforms to Requirements
Appearance (Form) Powder or Crystals
Infrared spectrum Conforms to Structure
Carbon 57.1 - 70.5%
Nitrogen 13.3 - 16.5%
GC (area %) ≥89.5%
Loss on Drying ≤10.5%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR