Bismut ≥99,9999% (Metall-Basis), Stäbe, Puratronic®, Länge 145 mm (5.71 in), Ø 10 mm (0.39 in)
Lieferant: Thermo Fisher Scientific
Synonyme:
Wismut
47229.KF
47229.KFEA
660
EUR
47229.KF
Bismut ≥99,9999% (Metall-Basis), Stäbe, Puratronic®, Länge 145 mm (5.71 in), Ø 10 mm (0.39 in)
Bismut
Purity: 99.9999%
CAS No.: 7440-69-9
Physical Form: ≈7.5g/cm
Odour: Odourless
MDL No.: MFCD00134033
Formel:
Bi Lagertemperatur: Raumtemperatur |
MDL:
MFCD00134033 CAS-Nummer: 7440-69-9 EINECS: 231-177-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...