D(+)-Glucose

Lieferant: Merck

Synonyme: Dextrose, Traubenzucker

47249
47249.EA 43.2 EUR
47249.
D(+)-Glucose
D(+)-Glucose
Formel: C₆H₁₂O₆
Molecular Weight: 180,16 g/mol
Schmelzpunkt: 146 °C
Dichte: 1,54 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00148912
CAS-Nummer: 50-99-7
EINECS: 200-075-1
Merck Index: 14,04459
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f93c10605c14be527901ce2420e2584a03e0a2fe&vid=f93c10605c14be527901ce2420e2584a03e0a2fe&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR