1,4,7-Triazacyclononan 90%

Lieferant: Thermo Scientific
Gefahr

Synonyme: 1,4,7-Triazonane, TACN

472520010
472520010.EA 223 EUR
472520010.
1,4,7-Triazacyclononan 90%
1,4,7-Triazacyclononane
Formel: C₆H₁₅N₃
Molecular Weight: 129,2 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00012358
CAS-Nummer: 4730-54-5
UN: 3259
ADR: 8,II

Bestellen

Spezifikation Testergebnisse

Appearance white to yellow low melting solid or liquid
Infrared spectrum Conforms
GC ≥88 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR