Acalabrutinib 98%

Lieferant: Thermo Scientific
Achtung

Synonyme: ACP-196, (S)-4-(8-amino-3-(1-(but-2-ynoyl)pyrrolidin-2-yl)imidazo[1,5-a]pyrazin-1-yl)-N-(pyridin-2-yl)benzamide

472531000
472531000.EA 280 EUR
472531000.
Acalabrutinib 98%
Acalabrutinib
Formel: C₂₆H₂₃N₇O₂
Molecular Weight: 465,52 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD29472294
CAS-Nummer: 1420477-60-6

Bestellen

Spezifikation Testergebnisse

Appearance white to almost white powder
Infrared spectrum Conforms
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR