Organic reference standards, Sugar Alcohol Kit, (individually packaged in quantities indicated), analytical standard, Supelco®
Lieferant: Merck
47266
47266.EA
295
EUR
47266.
Organic reference standards, Sugar Alcohol Kit, (individually packaged in quantities indicated), analytical standard, Supelco®
Standards
Organische Standards
Kit components:
D-(+)Arabitol 500 mg; Dulcitol (Galactitol) 500 mg; iso-Erythritol 500 mg; Glycerol 500 mg; Maltitol 500 mg; D-Mannitol 500 mg; Ribitol (Adonitol) 500 mg; D-Sorbitol 500 mg; Xylitol 500 mg.