Ethyl viologen diiodide 99%, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY

Synonyme: 1,1'-Diethyl-4,4'-bipyridinium diiodide

M-Clarity™ Qualitätsstufe = MQ100

472662-25G 472662-5G
472662-25GEA 381 EUR
472662-25G 472662-5G
Ethyl viologen diiodide 99%, Sigma-Aldrich®
Ethyl viologen diiodide

Ethyl viologen diiodide serves as an electrolyte to conduct cyclic voltammetry to investigate the kinetic process of the redox targeting reactions.

Formel: C₁₄H₁₈I₂N₂
Schmelzpunkt: 275 °C (Zers., Lit.)
MDL: MFCD00799539
CAS-Nummer: 1983-61-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=900e6fe9929fcab1945c53015abf1210e7cc5712&vid=3b5cafe2c56c1707d64b1541242e37e0bca8202f&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Appearance Orange to red powder, crystals, crystalline Powder and/or chunks
Infrared spectrum Conforms to structure
Elemental analysis Carbon 35.4%-36.5%, Nitrogen 5.8%- 6.2%
Titration 98.5%-101.5% (with silver nitrate)
High pressure liquid 98.5% (minimum)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR