4-((4-(Mesitylamino)pyrimidin-2-yl)amino)benzonitrile 98%

Lieferant: Thermo Scientific
Achtung

Synonyme: Dapivirine

472681000
472681000.EA 138 EUR
472681000.
4-((4-(Mesitylamino)pyrimidin-2-yl)amino)benzonitrile 98%
4-((4-(Mesitylamino)pyrimidin-2-yl)amino)benzonitrile
Formel: C₂₀H₁₉N₅
Molecular Weight: 329,39 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD09833899
CAS-Nummer: 244767-67-7

Bestellen

Spezifikation Testergebnisse

Appearance white to almost white powder
Infrared spectrum Conforms
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR