Letermovir 98%

Lieferant: Thermo Scientific
Achtung

Synonyme: (S)-2-(8-Fluoro-3-(2-methoxy-5-(trifluoromethyl)phenyl)-2-(4-(3-methoxyphenyl)piperazin-1-yl)-3,4-dihydroquinazolin-4-yl)acetic acid

472721000
472721000.EA 231 EUR
472721000.
Letermovir 98%
Letermovir
Formel: C₂₉H₂₈F₄N₄O₄
Molecular Weight: 572,55 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD22572725
CAS-Nummer: 917389-32-3

Bestellen

Spezifikation Testergebnisse

Appearance white to almost white powder
Infrared spectrum Conforms
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR