Specpure® Copper, Oil Based Standard Solution, Cu 1000 µg/g
Lieferant: Thermo Scientific
47276.18
47276.18EA
82.2
EUR
47276.18
Specpure® Copper, Oil Based Standard Solution, Cu 1000 µg/g
Standards
Kationischer Standards
Kupfer-Standards
Copper, Oil based standard solution, Specpure|r, Cu 1000^mg/g
Physical form: Liquid
MDL No.: MFCD00010965
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...