Cerium iron oxide (50:50 wt%) 20% in Wasser, Nanopartikel-Dispersion <5.0 nm APS NW, niedriger pH
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
47283.22
47283.36
47283.22EA
218
EUR
47283.22
47283.36
Cerium iron oxide (50:50 wt%) 20% in Wasser, Nanopartikel-Dispersion <5.0 nm APS NW, niedriger pH
Cerium iron oxide
Molecular Formula: CeO2:Fe2O3; =50:50 wt%
Physical Form: Liquid
Density: 1.23
Appearance: Red-brown
Odour: Petroleum-like
MDL No.: MFCD02687051
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...