AmberChrom™ chloride form 1x2 100-200 mesh

Lieferant: Thermo Fisher Scientific

473050025 473051000 473055000
473050025.EA 1317 EUR
473050025. 473051000. 473055000.
AmberChrom™ chloride form 1x2 100-200 mesh
AmberChrom™ chloride form
Formel: (C₁₀H₁₂·C₁₀H₁₀·C₈H₈·C₃H₉N)x
Lagertemperatur: Raumtemperatur
MDL: MFCD00146763
CAS-Nummer: 69011-19-4

Bestellen

Spezifikation Testergebnisse

Appearance white to yellow beads
Total exchange capacity ≥0.6 eq/l (Cl form)
Water retention capacity 65 to 75 % (Cl form)
Particle size 100 to 200 mesh


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR