Delavirdine mesylate 98%

Lieferant: Thermo Scientific

Synonyme: N-(2-(4-(3-(Isopropylamino)pyridin-2-yl)piperazine-1-carbonyl)-1H-indol-5-yl)methanesulphonamide methanesulphonate

473101000
473101000.EA 248 EUR
473101000.
Delavirdine mesylate 98%
Delavirdine mesylate
Formel: C₂₃H₃₂N₆O₆S₂
Molecular Weight: 552,67 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00866948
CAS-Nummer: 147221-93-0

Bestellen

Spezifikation Testergebnisse

Appearance white to light beige powder
Infrared spectrum Conforms
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR