Rhoifolin 97%
Lieferant: Thermo Scientific
Synonyme:
Apigenin-7-O-neohesperidoside, 7-(((2S,3R,4S,5S,6R)-4,5-Dihydroxy-6-(hydroxymethyl)-3-(((2S,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyltetrahydro-2H-pyran-2-yl)oxy)tetrahydro-2H-pyran-2-yl)oxy)-5-hydroxy-2-(4-hydroxyphenyl)-4H-chromen-4-one
473170500
473170500.EA
531
EUR
473170500.
Rhoifolin 97%
Rhoifolin
Formel:
C₂₇H₃₀O₁₄ Molecular Weight: 578,519 g/mol Lagertemperatur: Gefrierschrank |
MDL:
MFCD00016773 CAS-Nummer: 17306-46-6 EINECS: 241-335-4 |
Spezifikation Testergebnisse
Appearance | yellow powder |
Infrared spectrum | Conforms |
HPLC | ≥96 % |
Optical rotation | -90 to -110 (30°C, c=0.25, Methanol) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...