Alcaftadine 98+%
Lieferant: Thermo Scientific
Synonyme:
11-(1-Methylpiperidin-4-ylidene)-6,11-dihydro-5H-benzo[d]imidazo[1,2-a]azepine-3-carbaldehyde, 6,11-Dihydro-11-(1-methyl-4-piperidinylidene)-5H-imidazo[2,1-b][3]benzazepine-3-carboxaldehyde
473191000
473191000.EA
388
EUR
473191000.
Alcaftadine 98+%
Alcaftadine
Formel:
C₁₉H₂₁N₃O Molecular Weight: 307,40 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD09954106 CAS-Nummer: 147084-10-4 |
Spezifikation Testergebnisse
Appearance | white to light brown powder to crystals |
Infrared spectrum | Conforms |
HPLC | ≥98.0 % |
Melting point | 172°C to 175°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...