MK-4827 97%

Lieferant: Thermo Scientific
Gefahr

Synonyme: (S)-2-(4-(Piperidin-3-yl)phenyl)-2H-indazole-7-carboxamide, Niraparib

473221000
473221000.EA 273 EUR
473221000.
MK-4827 97%
MK-4827
Formel: C₁₉H₂₀N₄O
Molecular Weight: 320,39 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD17779309
CAS-Nummer: 1038915-60-4

Bestellen

Spezifikation Testergebnisse

Appearance white to light beige powder
Infrared spectrum Conforms
HPLC ≥96.0 %
Melting point 187.0°C to 202.0°C
Water ≤1.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR