[6]-Gingerol 97%

Lieferant: Thermo Scientific
Gefahr

Synonyme: (S)-5-Hydroxy-1-(4-hydroxy-3-methoxyphenyl)decan-3-one, 1-(4ʹ-Hydroxy-3ʹ-methoxyphenyl)-5-hydroxy-3-decanone, 6-Gingerol, (S)-5-Hydroxy-1-(4-hydroxy-3-methoxyphenyl)-3-decanone, Gingerol

473330500
473330500.EA 235 EUR
473330500.
[6]-Gingerol 97%
[6]-Gingerol
Formel: C₁₇H₂₆O₄
Molecular Weight: 294,4 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00210507
CAS-Nummer: 23513-14-6
UN: 2811
ADR: 6.1,III
Merck Index: 13,04436

Bestellen

Spezifikation Testergebnisse

Appearance colorless or light yellow viscous liquid or solid
Infrared spectrum Conforms
HPLC ≥96 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR