Idazoxan hydrochloride 97%

Lieferant: Thermo Scientific
Gefahr

Synonyme: 2-(2,3-Dihydrobenzo[b][1,4]dioxin-2-yl)-4,5-dihydro-1H-imidazole hydrochloride

473371000
473371000.EA 96 EUR
473371000.
Idazoxan hydrochloride 97%
Idazoxan hydrochloride
Formel: C₁₁H₁₂N₂O·HCl
Molecular Weight: 240,69 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00069293
CAS-Nummer: 79944-56-2
UN: 2811
ADR: 6.1,III
Merck Index: 13,04912

Bestellen

Spezifikation Testergebnisse

Appearance white to almost white solid
Infrared spectrum Conforms
HPLC ≥96 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR