D,L-Anatabine 95%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: 1,2,3,6-Tetrahydro-2,3'-bipyridine, (R,S)-Anatabine

473380010 473381000
473380010.EA 538 EUR
473380010. 473381000.
D,L-Anatabine 95%
D,L-Anatabine
Formel: C₁₀H₁₂N₂
Molecular Weight: 160.22 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00078278
CAS-Nummer: 2743-90-0
UN: 2810
ADR: 6.1 III

Bestellen

Spezifikation Testergebnisse

Appearance colorless to light yellow to orange liquid
Infrared spectrum Conforms
HPLC ≥94.0 %
Water ≤2.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR