Dibenzyl toluene Gemisch aus Isomeren 97%
Lieferant: Thermo Fisher Scientific
473440010
473442500
473440010.EA
83.8
EUR
473440010.
473442500.
Dibenzyl toluene Gemisch aus Isomeren 97%
Dibenzyl toluene
Formel:
C₂₁H₂₀ |
MDL:
MFCD01761976 CAS-Nummer: 53585-53-8 |
Spezifikation Testergebnisse
Appearance | colorless liquid |
Infrared spectrum | Conforms |
HPLC | ≥96 % (total isomers) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...