Ice Crusher
Lieferant: SCOTSMAN
CRUSHMAN360
474-0029EA 2080 EUR
474-0029
Ice Crusher
Eiszerkleinerer
The Crushman 360 is manufactured from stainless steel.
  • Three texture settings, fine, medium or coarse
  • Converts two kilograms of supercube ice into crushed granules in twenty seconds