Laboratory Ice Crusher
Lieferant: WARING BLENDOR
IC20
474-0044EA 1510 EUR
474-0044
Laboratory Ice Crusher
Eiszerkleinerer
The ice crusher is portable, of compact design and especially suitable for countertop use. It has a shaded pole gear motor, 24 rpm. The special height adjustment feature enables the use of any size container up to a maximum of 230 mm high. This is ideal for dispensing ice directly into a blender container.
  • Convenient crushed ice
  • Easily maintained
  • Height adjustment allows direct filling of large containers
Feed chute opening:  38×57 mm
Production of 568 g of ice per minute.