Pentaerythritol tetrakis [3-(3,5-di-tert-butyl-4-hydroxyphenyl)propionate] 95%
Lieferant: Thermo Fisher Scientific
Synonyme:
Tetrakis[methylene-3-(3,5-di-tert-butyl-4-hydroxyphenyl)propionate]methane, Antioxidant 1010, 2,2-Bis(((3-(3,5-di-tert-butyl-4-hydroxyphenyl)propanoyl)oxy)methyl)propane-1,3-diyl bis(3-(3,5-di-tert-butyl-4-hydroxyphenyl)propanoate)
474561000
474562500
474561000.EA
27.1
EUR
474561000.
474562500.
Pentaerythritol tetrakis [3-(3,5-di-tert-butyl-4-hydroxyphenyl)propionate] 95%
Pentaerythritol tetrakis [3-(3,5-di-tert-butyl-4-hydroxyphenyl)propionate]
Formel:
C₇₃H₁₀₈O₁₂ Siedepunkt: 1006°C (760mmHg) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00059345 CAS-Nummer: 6683-19-8 EINECS: 229-722-6 |
Spezifikation Testergebnisse
Appearance | white to almost white powder or crystals |
Infrared spectrum | Conforms |
HPLC | ≥94 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...