Tetrahydro-2-pyrimidinone 98+%
Lieferant: Thermo Scientific
Synonyme:
Tetrahydro-2(1H)-pyrimidinone, N,N'-Trimethyleneurea, Propyleneurea
474570500
474570500.EA
69.4
EUR
474570500.
Tetrahydro-2-pyrimidinone 98+%
Tetrahydro-2-pyrimidinone
Formel:
C₄H₈N₂O Molecular Weight: 100,12 g/mol Siedepunkt: 339°C (760mmHg) Flammpunkt: 158°C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00014593 CAS-Nummer: 1852-17-1 EINECS: 217-443-2 |
Spezifikation Testergebnisse
Appearance | white powder or crystals |
Infrared spectrum | Conforms |
HPLC | ≥98.0 % |
Melting point | 264°C to 268°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...